accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 696-44-6

IUPAC Name: N,3-dimethylaniline
Synonyms: benzenamine, N,3-dimethyl-
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: FBGJJTQNZVNEQU-UHFFFAOYSA-N
SMILES: CNC1=CC=CC(C)=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results