accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 696-68-4

IUPAC Name: [(cyclopenta-2,4-dien-1-ylidene)methyl]dimethylamine
Synonyms: (cyclopenta-2,4-dien-1-ylidenemethyl)dimethylamine
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: UJXXVGXQTXQALL-UHFFFAOYSA-N
SMILES: CN(C)C=C1C=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)