accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 696-82-2

IUPAC Name: 2,4,6-trifluoropyrimidine
Synonyms: 2,4,6-trifluoropyrimidine
Molecular Weight (g/mol): 134.06
Molecular Formula: C4HF3N2
InChi Key: NTSYSQNAPGMSIH-UHFFFAOYSA-N
SMILES: FC1=CC(F)=NC(F)=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)