accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6960-20-9

IUPAC Name: 3-methyl-5-nitropyridine
Synonyms: 3-methyl-5-nitropyridine
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: OQWXZZCTJZOSGH-UHFFFAOYSA-N
SMILES: CC1=CN=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results