accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOH

CAS RN 69617-84-1

IUPAC Name: 4-(3-hydroxybutyl)phenol
Synonyms: rhododendrol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: SFUCGABQOMYVJW-UHFFFAOYNA-N
SMILES: CC(O)CCC1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results