accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3NH2HCl

CAS RN 69630-50-8

IUPAC Name: 1,4-dimethyl 2-aminobutanedioate hydrochloride
Synonyms: 1,4-dimethyl 2-aminobutanedioate hydrochloride
Molecular Weight (g/mol): 197.62
Molecular Formula: C6H12ClNO4
InChi Key: PNLXWGDXZOYUKB-UHFFFAOYNA-N
SMILES: Cl.COC(=O)CC(N)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)