accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOHN

CAS RN 6964-62-1

IUPAC Name: 2-methyl-7-[phenyl(phenylamino)methyl]quinolin-8-ol
Synonyms: 2-methyl-7-[phenyl(phenylamino)methyl]quinolin-8-ol
Molecular Weight (g/mol): 340.43
Molecular Formula: C23H20N2O
InChi Key: WEENRMPCSWFMTE-UHFFFAOYNA-N
SMILES: CC1=CC=C2C=CC(C(NC3=CC=CC=C3)C3=CC=CC=C3)=C(O)C2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results