CAS RN 6966-09-2
IUPAC Name:
2-[(piperidin-1-yl)methyl]cyclohexan-1-one hydrochloridyl
Synonyms:
pimeclone hydrochloridyl
Molecular Weight (g/mol):
230.76
Molecular Formula:
C12H21ClNO
InChi Key:
MFMOSYZXKAREGX-UHFFFAOYNA-N
SMILES:
[Cl].O=C1CCCCC1CN1CCCCC1
1
–
1
of
1
results
1
Promotions Available
2-(Piperidylmethyl)-1-cyclohexanone hydrochloride, 98%, Thermo Scientific™
2-(Piperidylmethyl)-1-cyclohexanone hydrochloride, 98%, Thermo Scientific™
CAS: 6966-09-2 MDL Number: MFCD00068799 InChI Key: MFMOSYZXKAREGX-UHFFFAOYNA-N Synonym: pimeclone hydrochloride,2-piperidylmethyl-1-cyclohexanone hydrochloride,karion respiratory stimulant,1-piperidinomethyl-2-cyclohexanone hydrochloride,2-1-piperidinylmethyl cyclohexanone hydrochloride,2-piperidin-1-ylmethyl cyclohexanone hydrochloride,cyclohexanone, 2-piperidinomethyl-, hydrochloride,2-piperidin-1-ylmethyl cyclohexanone,pimeclone hcl,2-n-piperidinomethyl cyclohexanone hydrochloride PubChem CID: 197860