accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CCH3SOOO

CAS RN 69669-44-9

IUPAC Name: sodium 4-(dodecan-3-yl)benzene-1-sulfonate
Synonyms: sodium 4-(dodecan-3-yl)benzenesulfonate
Molecular Weight (g/mol): 348.48
Molecular Formula: C18H29NaO3S
InChi Key: HOXWFOVSCUWIEH-UHFFFAOYNA-M
SMILES: [Na+].CCCCCCCCCC(CC)C1=CC=C(C=C1)S([O-])(=O)=O
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)