accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrHOO

CAS RN 6967-82-4

IUPAC Name: 2-bromo-5-methylbenzoic acid
Synonyms: 2-bromo-5-methylbenzoic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: ZXMISUUIYPFORW-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results