accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69676-65-9

IUPAC Name: 2-[2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)ethoxy]acetic acid
Synonyms: [2-(1,3-dioxoisoindol-2-yl)ethoxy]acetic acid
Molecular Weight (g/mol): 249.22
Molecular Formula: C12H11NO5
InChi Key: PHZYUQLIZKTSJE-UHFFFAOYSA-N
SMILES: OC(=O)COCCN1C(=O)C2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results