accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6968-76-9

IUPAC Name: 1-(3-methoxyphenyl)piperazine hydrochloridyl
Synonyms: 1-(3-methoxyphenyl)piperazine hydrochloridyl
Molecular Weight (g/mol): 227.71
Molecular Formula: C11H16ClN2O
InChi Key: UYZISVLMMYHXMS-UHFFFAOYSA-N
SMILES: [Cl].COC1=CC=CC(=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results