accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNN

CAS RN 6969-71-7

IUPAC Name: 2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one
Synonyms: 2H-[1,2,4]triazolo[4,3-a]pyridin-3-one
Molecular Weight (g/mol): 135.13
Molecular Formula: C6H5N3O
InChi Key: LJRXNXBFJXXRNQ-UHFFFAOYSA-N
SMILES: O=C1NN=C2C=CC=CN12
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)