CAS RN 6970-19-0
IUPAC Name:
3,4,5-triethoxybenzoic acid
Synonyms:
3,4,5-triethoxybenzoic acid
Molecular Weight (g/mol):
254.28
Molecular Formula:
C13H18O5
InChi Key:
YCENSLDYFDZUMO-UHFFFAOYSA-N
SMILES:
CCOC1=CC(=CC(OCC)=C1OCC)C(O)=O
1
–
1
of
1
results
1
Promotions Available
3,4,5-Triethoxybenzoic acid, 98+%
3,4,5-Triethoxybenzoic acid, 98+%
CAS: 6970-19-0 Molecular Formula: C13H18O5 Molecular Weight (g/mol): 254.282 MDL Number: MFCD00002505 InChI Key: YCENSLDYFDZUMO-UHFFFAOYSA-N Synonym: benzoic acid,3,4,5-triethoxy,benzoic acid, 3,4,5-triethoxy,3,4,5-triethoxy benzoic acid,acmc-1b720 PubChem CID: 81432 IUPAC Name: 3,4,5-triethoxybenzoic acid SMILES: CCOC1=CC(=CC(=C1OCC)OCC)C(=O)O