CAS RN 69725-37-7
IUPAC Name:
2,2,7-trimethyloctane-3,5-dione
Synonyms:
2,2,7-trimethyloctane-3,5-dione
Molecular Weight (g/mol):
184.28
Molecular Formula:
C11H20O2
InChi Key:
KEBPOWLGOOTMPK-UHFFFAOYSA-N
SMILES:
CC(C)CC(=O)CC(=O)C(C)(C)C
1
–
1
of
1
results
1
Promotions Available
2,2,7-Trimethyloctane-3,5-dione, 97%
2,2,7-Trimethyloctane-3,5-dione, 97%
CAS: 69725-37-7 Molecular Formula: C11H20O2 Molecular Weight (g/mol): 184.279 MDL Number: MFCD00511279 InChI Key: KEBPOWLGOOTMPK-UHFFFAOYSA-N PubChem CID: 112295 IUPAC Name: 2,2,7-trimethyloctane-3,5-dione SMILES: CC(C)CC(=O)CC(=O)C(C)(C)C