CAS RN 6974-77-2
IUPAC Name:
1-bromo-3-chloro-2-methylpropane
Synonyms:
1-bromo-3-chloro-2-methylpropane
Molecular Weight (g/mol):
171.46
Molecular Formula:
C4H8BrCl
InChi Key:
ZKDOQFPDSUOLGF-UHFFFAOYNA-N
SMILES:
CC(CCl)CBr
1
–
1
of
1
results
1
1-Bromo-3-chloro-2-methylpropane 98.0+%, TCI America™
CAS: 6974-77-2 Molecular Formula: C4H8BrCl Molecular Weight (g/mol): 171.462 MDL Number: MFCD00000955 InChI Key: ZKDOQFPDSUOLGF-UHFFFAOYSA-N PubChem CID: 95566 IUPAC Name: 1-bromo-3-chloro-2-methylpropane SMILES: CC(CCl)CBr