accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6975-44-6

IUPAC Name: ethyl 6-hydroxy-4-oxo-1,4-dihydropyridine-3-carboxylate
Synonyms: ethyl 6-hydroxy-4-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 183.16
Molecular Formula: C8H9NO4
InChi Key: QDHHLXABEXNRJX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC(O)=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results