accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 6975-71-9

IUPAC Name: 2-(cyclohex-1-en-1-yl)acetonitrile
Synonyms: 2-(cyclohex-1-en-1-yl)acetonitrile
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: OYEXEQFKIPJKJK-UHFFFAOYSA-N
SMILES: N#CCC1=CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results