accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNHOHO

CAS RN 6976-17-6

IUPAC Name: 4-(methylamino)butanoic acid hydrochloridyl
Synonyms: 4-(methylamino)butyric acid hydrochloridyl
Molecular Weight (g/mol): 152.6
Molecular Formula: C5H11ClNO2
InChi Key: GYSZPWFLNPONPB-UHFFFAOYSA-N
SMILES: [Cl].CNCCCC(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results