accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69770-20-3

ClOO

CAS RN 69770-20-3

IUPAC Name: 3-(4-chlorophenoxy)benzaldehyde
Synonyms: 3-(4-chlorophenoxy)benzaldehyde
Molecular Weight (g/mol): 232.66
Molecular Formula: C13H9ClO2
InChi Key: MYQFRCYBOOWGJQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(OC2=CC=CC(C=O)=C2)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results