accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 698-80-6

IUPAC Name: 4-(chloromethyl)-1,2-difluorobenzene
Synonyms: 4-(chloromethyl)-1,2-difluorobenzene
Molecular Weight (g/mol): 162.56
Molecular Formula: C7H5ClF2
InChi Key: VTGRVYJPIVZZGS-UHFFFAOYSA-N
SMILES: FC1=CC=C(CCl)C=C1F
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results