CAS RN 69815-49-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ L-(–)-Norepinephrine-(+)-bitartrate
α,β-Adrenergic agonist.