accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNOHCH3CH3NH2NOO

CAS RN 69826-43-3

IUPAC Name: 4-carbamoyl-1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-imidazol-3-ium-3-olate
Synonyms: 5-carbamoyl-3-hydroxy-2,2,4,4-tetramethylimidazol-1-ium-1-olate
Molecular Weight (g/mol): 201.23
Molecular Formula: C8H15N3O3
InChi Key: UTSZZCVFWPSWBZ-UHFFFAOYSA-N
SMILES: CC1(C)N(O)C(C)(C)[N+]([O-])=C1C(N)=O
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results