accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69843-13-6

IUPAC Name: 1-methyl-1H-pyrazol-4-amine
Synonyms: 1-methylpyrazol-4-amine
Molecular Weight (g/mol): 97.12
Molecular Formula: C4H7N3
InChi Key: LBGSWBJURUFGLR-UHFFFAOYSA-N
SMILES: CN1C=C(N)C=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)