accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)ONH2OONaNa

CAS RN 69847-45-6

IUPAC Name: disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate
Synonyms: L-tyrosine disodium salt
Molecular Weight (g/mol): 225.16
Molecular Formula: C9H9NNa2O3
InChi Key: ASIYFCYUCMQNGK-RIYJSUBBNA-L
SMILES: [Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)