accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69876-37-5

IUPAC Name: benzyl (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoate
Synonyms: benzyl (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoate|Z-TRP-OBZL
Molecular Weight (g/mol): 428.49
Molecular Formula: C26H24N2O4
InChi Key: UHYCVEDLXBEKPC-DEOSSOPVSA-N
SMILES: O=C(N[C@@H](CC1=CNC2=CC=CC=C12)C(=O)OCC1=CC=CC=C1)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)