accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69888-86-4

IUPAC Name: 2,5-dioxopyrrolidin-1-yl tetradecanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl tetradecanoate
Molecular Weight (g/mol): 325.45
Molecular Formula: C18H31NO4
InChi Key: NZEDHZOVUDEBGW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results