accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHHO

CAS RN 699-83-2

IUPAC Name: 1-(2,6-dihydroxyphenyl)ethan-1-one
Synonyms: 1-(2,6-dihydroxyphenyl)ethanone
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: YPTJKHVBDCRKNF-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(O)C=CC=C1O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (5)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)