accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 699012-94-7

IUPAC Name: disodium tetracyanoplatinumbis(ylium)
Synonyms: disodium tetracyanoplatinumbis(ylium)
Molecular Weight (g/mol): 345.13
Molecular Formula: C4N4Na2Pt
InChi Key: WOIFBVMIVVRIIE-UHFFFAOYSA-N
SMILES: [Na+].[Na+].N#C[Pt++](C#N)(C#N)C#N
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results