accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69938-76-7

IUPAC Name: 3-(dimethylamino)-1-{6-[(N',N'-dimethylhydrazinecarbonyl)amino]hexyl}urea
Synonyms: 3-(dimethylamino)-1-{6-[(N',N'-dimethylhydrazinecarbonyl)amino]hexyl}urea
Molecular Weight (g/mol): 288.4
Molecular Formula: C12H28N6O2
InChi Key: VETHREXFBVHLJJ-UHFFFAOYSA-N
SMILES: CN(C)NC(=O)NCCCCCCNC(=O)NN(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results