accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69954-66-1

HClH3CNCH3OHO

CAS RN 69954-66-1

IUPAC Name: hydrogen 4-(dimethylamino)butanoic acid chloride
Synonyms: hydrogen 4-(dimethylamino)butanoic acid chloride
Molecular Weight (g/mol): 167.63
Molecular Formula: C6H14ClNO2
InChi Key: RDTALXUBMCLWBB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results