accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6996-92-5

IUPAC Name: benzeneseleninic acid
Synonyms: phenyl-selensaeure
Molecular Weight (g/mol): 189.08
Molecular Formula: C6H6O2Se
InChi Key: WIHKGDVGLJJAMC-UHFFFAOYSA-N
SMILES: O[Se](=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results