accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONCH3NNOOO

CAS RN 69975-86-6

IUPAC Name: 7-[(1,3-dioxolan-2-yl)methyl]-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: doxofylline maxivent dioxyfilline doxofilina doxophylline ventax 多索茶碱 doxofyllinum ansimar
Molecular Weight (g/mol): 266.26
Molecular Formula: C11H14N4O4
InChi Key: HWXIGFIVGWUZAO-UHFFFAOYSA-N
SMILES: CN1C2=C(N(CC3OCCO3)C=N2)C(=O)N(C)C1=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)