accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70-00-8

IUPAC Name: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: trifluridine|trifluridina [inn-spanish]
Molecular Weight (g/mol): 296.2
Molecular Formula: C10H11F3N2O5
InChi Key: VSQQQLOSPVPRAZ-RRKCRQDMSA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C(=O)NC1=O)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)