CAS RN 70-11-1
2-Bromoacetophenone, 98%
CAS: 70-11-1 Molecular Formula: C8H7BrO Molecular Weight (g/mol): 199.047 MDL Number: MFCD00000195 InChI Key: LIGACIXOYTUXAW-UHFFFAOYSA-N Synonym: 2-bromoacetophenone,phenacyl bromide,2-bromo-1-phenylethan-1-one,bromoacetophenone,ethanone, 2-bromo-1-phenyl,bromomethyl phenyl ketone,alpha-bromoacetophenone,stauffer 4644,omega-bromoacetophenone,acetophenone, 2-bromo PubChem CID: 6259 ChEBI: CHEBI:51846 IUPAC Name: 2-bromo-1-phenylethanone SMILES: C1=CC=C(C=C1)C(=O)CBr
Phenacyl Bromide 98.0+%, TCI America™
CAS: 70-11-1 Molecular Formula: C8H7BrO Molecular Weight (g/mol): 199.047 MDL Number: MFCD00000195 InChI Key: LIGACIXOYTUXAW-UHFFFAOYSA-N Synonym: 2-bromoacetophenone,phenacyl bromide,2-bromo-1-phenylethan-1-one,bromoacetophenone,ethanone, 2-bromo-1-phenyl,bromomethyl phenyl ketone,alpha-bromoacetophenone,stauffer 4644,omega-bromoacetophenone,acetophenone, 2-bromo PubChem CID: 6259 ChEBI: CHEBI:51846 IUPAC Name: 2-bromo-1-phenylethanone SMILES: C1=CC=C(C=C1)C(=O)CBr
2-Bromoacetophenone, 98%
CAS: 70-11-1 Molecular Formula: C8H7BrO Molecular Weight (g/mol): 199.05 MDL Number: MFCD00000195 InChI Key: LIGACIXOYTUXAW-UHFFFAOYSA-N Synonym: 2-bromoacetophenone,phenacyl bromide,2-bromo-1-phenylethan-1-one,bromoacetophenone,ethanone, 2-bromo-1-phenyl,bromomethyl phenyl ketone,alpha-bromoacetophenone,stauffer 4644,omega-bromoacetophenone,acetophenone, 2-bromo PubChem CID: 6259 ChEBI: CHEBI:51846 IUPAC Name: 2-bromo-1-phenylethanone SMILES: C1=CC=C(C=C1)C(=O)CBr
Phenacyl Bromide 98.0+%, TCI America™
CAS: 70-11-1 Molecular Formula: C8H7BrO Molecular Weight (g/mol): 199.047 MDL Number: MFCD00000195 InChI Key: LIGACIXOYTUXAW-UHFFFAOYSA-N Synonym: 2-bromoacetophenone,phenacyl bromide,2-bromo-1-phenylethan-1-one,bromoacetophenone,ethanone, 2-bromo-1-phenyl,bromomethyl phenyl ketone,alpha-bromoacetophenone,stauffer 4644,omega-bromoacetophenone,acetophenone, 2-bromo PubChem CID: 6259 ChEBI: CHEBI:51846 IUPAC Name: 2-bromo-1-phenylethanone SMILES: C1=CC=C(C=C1)C(=O)CBr
Sigma Aldrich 2-Bromoacetophenone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Phenacyl Bromide 97.0+%, TCI America™
CAS: 70-11-1 Molecular Formula: C8H7BrO Molecular Weight (g/mol): 199.047 MDL Number: MFCD00000195 InChI Key: LIGACIXOYTUXAW-UHFFFAOYSA-N Synonym: 2-bromoacetophenone,phenacyl bromide,2-bromo-1-phenylethan-1-one,bromoacetophenone,ethanone, 2-bromo-1-phenyl,bromomethyl phenyl ketone,alpha-bromoacetophenone,stauffer 4644,omega-bromoacetophenone,acetophenone, 2-bromo PubChem CID: 6259 ChEBI: CHEBI:51846 IUPAC Name: 2-bromo-1-phenylethanone SMILES: C1=CC=C(C=C1)C(=O)CBr