accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70-23-5

IUPAC Name: ethyl 3-bromo-2-oxopropanoate
Synonyms: ethyl 3-bromo-2-oxopropanoate
Molecular Weight (g/mol): 195.01
Molecular Formula: C5H7BrO3
InChi Key: VICYTAYPKBLQFB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)CBr
Boiling Point 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)