accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 700-00-5

IUPAC Name: 9-methyl-9H-purin-6-amine
Synonyms: 9-methyladenine
Molecular Weight (g/mol): 149.16
Molecular Formula: C6H7N5
InChi Key: WRXCXOUDSPTXNX-UHFFFAOYSA-N
SMILES: CN1C=NC2=C(N)N=CN=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results