accessibility menu, dialog, popup

UserName

Viewing profile as user:
FNFFFF

CAS RN 700-16-3

IUPAC Name: pentafluoropyridine
Synonyms: pentafluoropyridine
Molecular Weight (g/mol): 169.05
Molecular Formula: C5F5N
InChi Key: XTGOWLIKIQLYRG-UHFFFAOYSA-N
SMILES: FC1=NC(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (4)