accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70024-40-7

IUPAC Name: hydrogen 6,7-dimethoxy-2-[4-(oxolane-2-carbonyl)piperazin-1-yl]quinazolin-4-amine dihydrate chloride
Synonyms: terazosin hydrochloride dihydrate terazosin monohydrochloride dihydrate
Molecular Weight (g/mol): 459.93
Molecular Formula: C19H30ClN5O6
InChi Key: NZMOFYDMGFQZLS-UHFFFAOYNA-N
SMILES: [H+].O.O.[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1CCCO1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)