accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70095-40-8

IUPAC Name: hexan-2-amine
Synonyms: hexan-2-amine
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: WGBBUURBHXLGFM-UHFFFAOYNA-N
SMILES: CCCCC(C)N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results