accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70097-65-3

IUPAC Name: 1-(benzyloxy)-4-(trifluoromethyl)benzene
Synonyms: 1-(benzyloxy)-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 252.24
Molecular Formula: C14H11F3O
InChi Key: SNRFAICCYVUTRG-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results