accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SOOBr

CAS RN 701-34-8

IUPAC Name: 4-bromobenzene-1-sulfonamide
Synonyms: 4-bromobenzenesulfonamide
Molecular Weight (g/mol): 236.08
Molecular Formula: C6H6BrNO2S
InChi Key: STYQHICBPYRHQK-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results