accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiClClCl

CAS RN 701-35-9

IUPAC Name: trichloro(4-methylphenyl)silane
Synonyms: trichloro(4-methylphenyl)silane
Molecular Weight (g/mol): 225.57
Molecular Formula: C7H7Cl3Si
InChi Key: WOMUGKOOLXQCTQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)[Si](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)