accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 701-45-1

IUPAC Name: 2-bromo-1-fluoro-4-nitrobenzene
Synonyms: 2-bromo-1-fluoro-4-nitrobenzene
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: FAWMTDSAMOCUAR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C(Br)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)