accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 701-64-4

IUPAC Name: disodium phenyl phosphate
Synonyms: disodium phenyl phosphate
Molecular Weight (g/mol): 218.06
Molecular Formula: C6H5Na2O4P
InChi Key: TYJOJLOWRIQYQM-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]P([O-])(=O)OC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results