accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 701-82-6

IUPAC Name: (3-hydroxyphenyl)urea
Synonyms: M-hydroxyphenylurea
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: IPRCBIWIPMJXIK-UHFFFAOYSA-N
SMILES: NC(=O)NC1=CC=CC(O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results