accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7012-37-5

IUPAC Name: 2,4,4'-trichloro-1,1'-biphenyl
Synonyms: 2,4,4'-trichlorobiphenyl
Molecular Weight (g/mol): 257.54
Molecular Formula: C12H7Cl3
InChi Key: BZTYNSQSZHARAZ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results