accessibility menu, dialog, popup

UserName

Viewing profile as user:
A(a)

CAS RN 70131-67-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Quantity 
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)