accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHON

CAS RN 70165-31-0

IUPAC Name: 6-cyanopyridine-3-carboxylic acid
Synonyms: 6-cyanopyridine-3-carboxylic acid
Molecular Weight (g/mol): 148.12
Molecular Formula: C7H4N2O2
InChi Key: WMHSQCDPPJRWIL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)