CAS RN 701947-53-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PIP3 Antagonist II, DM-PIT-1
The PIP3 Antagonist II, DM-PIT-1, also referenced under CAS 701947-53-7, controls the biological activity of PIP3.